siRNA Details
| Gene Symbol: Mettl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146247.1 | NP_666359.1 | 3, 4 | 546 | ||
| GenBank mRNA | AK089212.1 | - | 3, 4 | 561 | ||
| AK136614.1 | - | 3, 4 | 572 | |||
| BC024814.1 | - | 3, 4 | 546 | 5359405 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl22| Gene Symbol: Mettl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146247.1 | NP_666359.1 | 3, 4 | 546 | ||
| GenBank mRNA | AK089212.1 | - | 3, 4 | 561 | ||
| AK136614.1 | - | 3, 4 | 572 | |||
| BC024814.1 | - | 3, 4 | 546 | 5359405 | ||