siRNA Details
| Gene Symbol: Mkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153588.3 | NP_705816.2 | 1 | 279 | ||
| GenBank mRNA | AF532598.1 | - | 1 | 61 | ||
| AK158754.1 | - | 1 | 278 | |||
| AK220377.1 | - | 1 | 61 | |||
| BC131644.1 | - | 1 | 61 | 40110019 | ||
| BC131645.1 | - | 1 | 59 | |||
| CJ126058.1 | - | 1 | 281 | |||

Mkl2
Genomic Map