siRNA Details
| Gene Symbol: Rimbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033338.3 | NP_001028510.2 | 1 | 3256 | ||
| GenBank mRNA | AK133200.1 | - | 1 | 2407 | ||
| AK220410.1 | - | 1 | 1276 | |||
| BC141386.1 | - | 1 | 2400 | 9055999 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rimbp3| Gene Symbol: Rimbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033338.3 | NP_001028510.2 | 1 | 3256 | ||
| GenBank mRNA | AK133200.1 | - | 1 | 2407 | ||
| AK220410.1 | - | 1 | 1276 | |||
| BC141386.1 | - | 1 | 2400 | 9055999 | ||