siRNA Details
| Gene Symbol: Klhl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183390.3 | NP_899246.2 | 5 | 1286 | ||
| GenBank mRNA | AY350736.1 | - | 5 | 1214 | ||
| BC100568.1 | - | 5 | 1273 | 30073940 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl6| Gene Symbol: Klhl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183390.3 | NP_899246.2 | 5 | 1286 | ||
| GenBank mRNA | AY350736.1 | - | 5 | 1214 | ||
| BC100568.1 | - | 5 | 1273 | 30073940 | ||