siRNA Details
| Gene Symbol: Ostn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198112.2 | NP_932780.1 | 3 | 358 | ||
| GenBank mRNA | AY395730.1 | - | 3 | 358 | ||
| AY573932.1 | - | 3 | 510 | |||
| BC117063.1 | - | 3 | 346 | 40126382 | ||
| BC117089.1 | - | 3 | 346 | 40126408 | ||

Ostn
Genomic Map