siRNA Details
| Gene Symbol: Lmln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172823.2 | NP_766411.1 | 6, 7 | 734 | ||
| GenBank mRNA | AK030463.1 | - | 6, 7 | 735 | ||
| BC111899.1 | - | 6, 7 | 735 | 40062192 | ||
| BC112428.1 | - | 6, 7 | 735 | 40062196 | ||

Lmln
Genomic Map