siRNA Details
| Gene Symbol: Gpr156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153394.2 | NP_700443.2 | 2, 3 | 567 | ||
| GenBank mRNA | AF488740.1 | - | 1, 2 | 91 | ||
| AK141990.1 | - | 2, 3 | 612 | |||
| BC059227.1 | - | 2, 3 | 567 | 6400663 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr156| Gene Symbol: Gpr156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153394.2 | NP_700443.2 | 2, 3 | 567 | ||
| GenBank mRNA | AF488740.1 | - | 1, 2 | 91 | ||
| AK141990.1 | - | 2, 3 | 612 | |||
| BC059227.1 | - | 2, 3 | 567 | 6400663 | ||