siRNA Details
| Gene Symbol: Cadm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145977.1 | NP_001139449.1 | 7 | 1507 | ||
| NM_178721.4 | NP_848836.2 | 8 | 1534 | |||
| GenBank mRNA | AK033973.1 | - | 7 | 1134 | ||
| AK038842.1 | - | 8 | 1513 | |||
| AK043760.1 | - | 8 | 1517 | |||
| AK046800.1 | - | 7 | 1410 | |||
| AK163636.1 | - | 8 | 1511 | |||
| AK165030.1 | - | 8 | 1531 | |||
| BC139401.1 | - | 8 | 1103 | 8862423 | ||
| BC139402.1 | - | 8 | 1103 | 8862424 | ||

Cadm2
Genomic Map