siRNA Details
Gene Symbol: Krtap24-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163141.1 | NP_001156613.1 | 1 | 601 | ||
GenBank mRNA | AK037168.1 | - | 1 | 602 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Krtap24-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163141.1 | NP_001156613.1 | 1 | 601 | ||
GenBank mRNA | AK037168.1 | - | 1 | 602 |