siRNA Details
| Gene Symbol: Gm4944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205095.1 | NP_001192024.1 | 5 | 408 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4944| Gene Symbol: Gm4944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205095.1 | NP_001192024.1 | 5 | 408 | ||