siRNA Details
| Gene Symbol: Zfp799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177359.5 | NP_796333.3 | 4 | 1094 | ||
| GenBank mRNA | AK031686.1 | - | 4 | 1082 | ||
| AK083640.1 | - | 4 | 1071 | |||
| AK137037.1 | - | 4 | 1071 | |||
| BC096552.1 | - | 4 | 1044 | 30618827 | ||

Zfp799
Genomic Map