siRNA Details
| Gene Symbol: St6gal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172829.2 | NP_766417.1 | 2 | 261 | ||
| GenBank mRNA | AK054230.1 | - | 2 | 597 | ||
| AK082566.1 | - | 2 | 262 | |||
| BC125586.1 | - | 1 | 131 | 40130101 | ||
| BC138142.1 | - | 1 | 131 | 8861161 | ||

St6gal2
Genomic Map