siRNA Details
| Gene Symbol: Zfp119b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146249.4 | NP_666361.1 | 4 | 1480 | ||
| GenBank mRNA | BC031441.1 | - | 4 | 1480 | 3985007 | |
| BC094043.1 | - | 1669 | 5067417 | |||
| BC137918.1 | - | 4 | 1463 | 8860937 | ||
| BC137919.1 | - | 4 | 1463 | 8860938 | ||

Zfp119b
Genomic Map