siRNA Details
| Gene Symbol: Fsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183178.2 | NP_899001.1 | 8 | 858 | ||
| GenBank mRNA | AK140132.1 | - | 8 | 892 | ||
| AK144257.1 | - | 8 | 793 | |||
| BC055106.1 | - | 8 | 858 | 6532001 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fsd1| Gene Symbol: Fsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183178.2 | NP_899001.1 | 8 | 858 | ||
| GenBank mRNA | AK140132.1 | - | 8 | 892 | ||
| AK144257.1 | - | 8 | 793 | |||
| BC055106.1 | - | 8 | 858 | 6532001 | ||