siRNA Details
| Gene Symbol: Thada | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183021.3 | NP_898842.2 | 6 | 647 | ||
| GenBank mRNA | AK044550.1 | - | 6 | 600 | ||
| AK154078.1 | - | 6 | 645 | |||
| EF222207.1 | - | 6 | 555 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thada| Gene Symbol: Thada | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183021.3 | NP_898842.2 | 6 | 647 | ||
| GenBank mRNA | AK044550.1 | - | 6 | 600 | ||
| AK154078.1 | - | 6 | 645 | |||
| EF222207.1 | - | 6 | 555 | |||