siRNA Details
| Gene Symbol: 9430020K01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081963.1 | NP_001075432.1 | 3 | 4025 | ||
| GenBank mRNA | AK034658.1 | - | 1 | 1081 | ||
| AK220394.1 | - | 1 | 2182 | |||
| BC144919.1 | - | 2 | 3929 | 9053467 | ||

9430020K01Rik
Genomic Map