siRNA Details
| Gene Symbol: Ythdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163013.1 | NP_001156485.1 | 27 | 3934 | ||
| GenBank mRNA | AK140113.1 | - | 1 | 44 | ||
| BC037178.1 | - | 14 | 1930 | |||
| BC040772.1 | - | 8 | 1221 | |||
| BC072654.1 | - | 22 | 2978 | |||
| BC138263.1 | - | 27 | 3934 | 8861282 | ||
| BC171951.1 | - | 27 | 3934 | 9054237 | ||

Ythdc2
Genomic Map