siRNA Details
| Gene Symbol: Ccdc112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160399.1 | NP_001153871.1 | 6 | 815 | ||
| GenBank mRNA | AA241279.1 | - | 3 | 291 | ||
| AK043959.1 | - | 1 | 1566 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc112| Gene Symbol: Ccdc112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160399.1 | NP_001153871.1 | 6 | 815 | ||
| GenBank mRNA | AA241279.1 | - | 3 | 291 | ||
| AK043959.1 | - | 1 | 1566 | |||