siRNA Details
| Gene Symbol: Ccdc112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160399.1 | NP_001153871.1 | 7, 8 | 1457 | ||
| GenBank mRNA | BC119623.1 | - | 2, 3 | 81 | ||
| BC119624.1 | - | 2, 3 | 48 | |||
| DV656314.1 | - | 1, 2 | 356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc112| Gene Symbol: Ccdc112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160399.1 | NP_001153871.1 | 7, 8 | 1457 | ||
| GenBank mRNA | BC119623.1 | - | 2, 3 | 81 | ||
| BC119624.1 | - | 2, 3 | 48 | |||
| DV656314.1 | - | 1, 2 | 356 | |||