siRNA Details
| Gene Symbol: Adnp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175028.1 | NP_778193.1 | 4 | 926 | ||
| GenBank mRNA | AB093268.1 | - | 4 | 926 | ||
| BC044898.1 | - | 4 | 831 | |||
| BC047352.1 | - | 4 | 831 | |||
| BC064672.1 | - | 2 | 699 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adnp2| Gene Symbol: Adnp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175028.1 | NP_778193.1 | 4 | 926 | ||
| GenBank mRNA | AB093268.1 | - | 4 | 926 | ||
| BC044898.1 | - | 4 | 831 | |||
| BC047352.1 | - | 4 | 831 | |||
| BC064672.1 | - | 2 | 699 | |||