siRNA Details
| Gene Symbol: Kcng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190373.1 | NP_001177302.1 | 4 | 1637 | ||
| GenBank mRNA | AK158780.1 | - | 1 | 615 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcng2| Gene Symbol: Kcng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190373.1 | NP_001177302.1 | 4 | 1637 | ||
| GenBank mRNA | AK158780.1 | - | 1 | 615 | ||