siRNA Details
| Gene Symbol: Cdc42bpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033342.1 | NP_001028514.1 | 31 | 4240 | ||
| GenBank mRNA | AK133191.1 | - | 8 | 1312 | ||
| BC046418.1 | - | 14 | 1888 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc42bpg| Gene Symbol: Cdc42bpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033342.1 | NP_001028514.1 | 31 | 4240 | ||
| GenBank mRNA | AK133191.1 | - | 8 | 1312 | ||
| BC046418.1 | - | 14 | 1888 | |||