siRNA Details
| Gene Symbol: Ranbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177721.4 | NP_808389.2 | 1 | 985 | ||
| GenBank mRNA | AK044143.1 | - | 1 | 970 | ||
| BC056759.1 | - | 1 | 964 | 6518485 | ||
| BC139395.1 | - | 1 | 985 | 8862417 | ||
| BC139396.1 | - | 1 | 985 | 8862418 | ||

Ranbp6
Genomic Map