siRNA Details
| Gene Symbol: Slco5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172841.2 | NP_766429.2 | 6 | 2363 | ||
| GenBank mRNA | AK041736.1 | - | 6 | 2364 | ||
| BC117943.1 | - | 5 | 1943 | 40097907 | ||
| BC117944.1 | - | 4 | 1765 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slco5a1| Gene Symbol: Slco5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172841.2 | NP_766429.2 | 6 | 2363 | ||
| GenBank mRNA | AK041736.1 | - | 6 | 2364 | ||
| BC117943.1 | - | 5 | 1943 | 40097907 | ||
| BC117944.1 | - | 4 | 1765 | |||