siRNA Details
| Gene Symbol: Plekha6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160268.1 | NP_001153740.1 | 10 | 1773 | ||
| NM_182930.2 | NP_891846.1 | 13 | 1935 | |||
| GenBank mRNA | AK149273.1 | - | 4 | 1128 | ||
| AK173074.1 | - | 10 | 1744 | |||
| BC038061.1 | - | 9 | 1710 | |||
| BC054547.1 | - | 13 | 1926 | 5709381 | ||

Plekha6
Genomic Map