siRNA Details
| Gene Symbol: Teddm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178243.3 | NP_839974.2 | 1 | 346 | ||
| GenBank mRNA | AJ554213.1 | - | 1 | 303 | ||
| AK136655.1 | - | 1 | 348 | |||
| BC120491.1 | - | 1 | 300 | 40129414 | ||
| BC131905.1 | - | 1 | 302 | 40130182 | ||
| DQ402363.1 | - | 1 | 346 | |||

Teddm2
Genomic Map