siRNA Details
| Gene Symbol: Teddm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178244.3 | NP_839975.2 | 1 | 351 | ||
| GenBank mRNA | AJ554212.1 | - | 1 | 205 | ||
| BC109332.2 | - | 1 | 204 | |||
| BC141344.1 | - | 1 | 254 | 9055956 | ||
| BC141345.1 | - | 1 | 255 | 9055957 | ||

Teddm1a
Genomic Map