siRNA Details
| Gene Symbol: Tor1aip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172843.4 | NP_766431.3 | 7 | 1878 | ||
| GenBank mRNA | AK038024.1 | - | 7 | 2140 | ||
| AK080959.1 | - | 5 | 1157 | |||
| BC066790.1 | - | 5 | 1084 | 30099444 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tor1aip2| Gene Symbol: Tor1aip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172843.4 | NP_766431.3 | 7 | 1878 | ||
| GenBank mRNA | AK038024.1 | - | 7 | 2140 | ||
| AK080959.1 | - | 5 | 1157 | |||
| BC066790.1 | - | 5 | 1084 | 30099444 | ||