siRNA Details
| Gene Symbol: Brinp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207583.2 | NP_997466.2 | 3 | 1108 | ||
| GenBank mRNA | AK032297.1 | - | 3 | 1112 | ||
| AK045801.1 | - | 3 | 635 | |||
| AK122549.1 | - | 3 | 1174 | |||
| BC067038.1 | - | 4 | 754 | |||
| BC076572.1 | - | 3 | 1108 | 30363878 | ||

Brinp2
Genomic Map