siRNA Details
| Gene Symbol: Vsig8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177723.4 | NP_808391.2 | 4 | 618 | ||
| GenBank mRNA | AK037228.1 | - | 4 | 619 | ||
| BC064106.1 | - | 3 | 337 | 1528324 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vsig8| Gene Symbol: Vsig8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177723.4 | NP_808391.2 | 4 | 618 | ||
| GenBank mRNA | AK037228.1 | - | 4 | 619 | ||
| BC064106.1 | - | 3 | 337 | 1528324 | ||