siRNA Details
| Gene Symbol: Carf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285463.1 | NP_001272392.1 | 12 | 1287 | ||
| NM_001285473.1 | NP_001272402.1 | 8 | 877 | |||
| NM_139150.5 | NP_631889.1 | 10 | 1160 | |||
| GenBank mRNA | AB221600.1 | - | 9 | 943 | ||
| AF454947.1 | - | 10 | 1129 | |||
| AK044738.1 | - | 12 | 1288 | |||
| BC138644.1 | - | 9 | 1066 | 8861664 | ||
| BC138645.1 | - | 9 | 1066 | 8861665 | ||
| BC145350.1 | - | 9 | 1062 | |||
| EU170502.1 | - | 8 | 858 | |||

Carf
Genomic Map