siRNA Details
| Gene Symbol: Carf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285463.1 | NP_001272392.1 | 17, 18 | 2308 | ||
| NM_001285473.1 | NP_001272402.1 | 13, 14 | 1898 | |||
| NM_139150.5 | NP_631889.1 | 15, 16 | 2181 | |||
| GenBank mRNA | AB221600.1 | - | 14, 15 | 1964 | ||
| AF454947.1 | - | 15, 16 | 2150 | |||
| AK044738.1 | - | 17, 18 | 2309 | |||
| BC138644.1 | - | 14, 15 | 2087 | 8861664 | ||
| BC138645.1 | - | 14, 15 | 2087 | 8861665 | ||
| BC145350.1 | - | 14, 15 | 2077 | |||
| EU170502.1 | - | 13, 14 | 1879 | |||
| EU170503.1 | - | 11, 12 | 1614 | |||

Carf
Genomic Map