siRNA Details
| Gene Symbol: Plekhm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039493.1 | NP_001034582.1 | 4 | 2026 | ||
| GenBank mRNA | AK034959.1 | - | 4 | 1948 | ||
| AK172220.1 | - | 3 | 1119 | |||
| BC063098.1 | - | 2026 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhm3| Gene Symbol: Plekhm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039493.1 | NP_001034582.1 | 4 | 2026 | ||
| GenBank mRNA | AK034959.1 | - | 4 | 1948 | ||
| AK172220.1 | - | 3 | 1119 | |||
| BC063098.1 | - | 2026 | ||||