siRNA Details
Gene Symbol: Fam124b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173425.3 | NP_775601.1 | 2 | 1574 | ||
GenBank mRNA | AK043867.1 | - | 2 | 1575 | ||
BC150872.1 | - | 2 | 1459 | 9087783 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam124b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173425.3 | NP_775601.1 | 2 | 1574 | ||
GenBank mRNA | AK043867.1 | - | 2 | 1575 | ||
BC150872.1 | - | 2 | 1459 | 9087783 |