siRNA Details
| Gene Symbol: Nyap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172849.3 | NP_766437.2 | 3 | 891 | ||
| GenBank mRNA | AB429290.1 | - | 3 | 382 | ||
| AK034747.1 | - | 4 | 906 | |||
| AK043512.1 | - | 3 | 626 | |||
| AK049778.1 | - | 4 | 900 | |||
| AK137094.1 | - | 3 | 888 | |||
| BC082310.1 | - | 3 | 891 | 30355797 | ||

Nyap2
Genomic Map