siRNA Details
| Gene Symbol: Neu4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173772.3 | NP_776133.1 | 4 | 1369 | ||
| GenBank mRNA | AB510404.1 | - | 5 | 1350 | ||
| AK034236.1 | - | 4 | 1340 | |||
| AY258421.1 | - | 4 | 1282 | |||
| BC070438.1 | - | 4 | 1369 | 6842155 | ||

Neu4
Genomic Map