siRNA Details
| Gene Symbol: Serpinb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172852.3 | NP_766440.2 | 7 | 785 | ||
| GenBank mRNA | AK030670.1 | - | 7 | 787 | ||
| BC117990.1 | - | 7 | 775 | 40104505 | ||
| BC117991.1 | - | 7 | 722 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpinb13| Gene Symbol: Serpinb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172852.3 | NP_766440.2 | 7 | 785 | ||
| GenBank mRNA | AK030670.1 | - | 7 | 787 | ||
| BC117990.1 | - | 7 | 775 | 40104505 | ||
| BC117991.1 | - | 7 | 722 | |||