siRNA Details
| Gene Symbol: Gpr158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004761.1 | NP_001004761.1 | 10 | 2721 | ||
| GenBank mRNA | AB093287.1 | - | 10 | 1826 | ||
| AK083234.1 | - | 10 | 2747 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr158| Gene Symbol: Gpr158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004761.1 | NP_001004761.1 | 10 | 2721 | ||
| GenBank mRNA | AB093287.1 | - | 10 | 1826 | ||
| AK083234.1 | - | 10 | 2747 | |||