siRNA Details
| Gene Symbol: Ralgps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290570.1 | NP_001277499.1 | 6, 7 | 726 | ||
| NM_001290572.1 | NP_001277501.1 | 7, 8 | 777 | |||
| NM_175211.5 | NP_780420.1 | 7, 8 | 777 | |||
| GenBank mRNA | AK076767.1 | - | 7, 8 | 762 | ||
| AK129123.1 | - | 6, 7 | 693 | |||
| AK170248.1 | - | 7, 8 | 724 | |||

Ralgps1
Genomic Map