siRNA Details
| Gene Symbol: Cers6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172856.3 | NP_766444.1 | 7 | 746 | ||
| GenBank mRNA | AK028849.1 | - | 7 | 747 | ||
| AK135798.1 | - | 7 | 848 | |||
| AK151848.1 | - | 7 | 835 | |||
| AK152417.1 | - | 7 | 816 | |||
| BC057629.1 | - | 7 | 789 | 4224547 | ||

Cers6
Genomic Map