siRNA Details
| Gene Symbol: Harbi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178724.4 | NP_848839.2 | 2 | 553 | ||
| GenBank mRNA | AK041747.1 | - | 1 | 136 | ||
| AK045343.1 | - | 2 | 543 | |||
| AK080671.1 | - | 2 | 582 | |||
| AK084226.1 | - | 2 | 587 | |||
| AK147045.1 | - | 2 | 593 | |||
| AK214529.1 | - | 1 | 78 | |||
| BC049253.1 | - | 1 | 58 | |||
| BC094315.1 | - | 2 | 553 | 5716740 | ||

Harbi1
Genomic Map