siRNA Details
| Gene Symbol: D430041D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033347.2 | NP_001028519.2 | 6 | 1825 | ||
| GenBank mRNA | CX239849.1 | - | 3 | 401 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D430041D05Rik| Gene Symbol: D430041D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033347.2 | NP_001028519.2 | 6 | 1825 | ||
| GenBank mRNA | CX239849.1 | - | 3 | 401 | ||