siRNA Details
| Gene Symbol: D430041D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033347.2 | NP_001028519.2 | 14 | 2960 | ||
| GenBank mRNA | AK047363.1 | - | 8 | 1075 | ||
| AK052518.1 | - | 5 | 863 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D430041D05Rik| Gene Symbol: D430041D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033347.2 | NP_001028519.2 | 14 | 2960 | ||
| GenBank mRNA | AK047363.1 | - | 8 | 1075 | ||
| AK052518.1 | - | 5 | 863 | |||