siRNA Details
| Gene Symbol: Slc5a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003915.2 | NP_001003915.1 | 12 | 1710 | ||
| NM_001177624.1 | NP_001171095.1 | 12 | 1698 | |||
| GenBank mRNA | AF529222.1 | - | 12 | 1718 | ||
| AK085356.1 | - | 12 | 1693 | |||
| AK128951.1 | - | 12 | 1690 | |||
| AK144004.1 | - | 12 | 1631 | |||
| AK165419.1 | - | 12 | 1691 | |||
| AY964639.1 | - | 12 | 1401 | |||
| BC121791.1 | - | 12 | 1394 | |||
| BC141255.1 | - | 12 | 1626 | 9055867 | ||

Slc5a12
Genomic Map