siRNA Details
| Gene Symbol: Atp8b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080944.3 | NP_001074413.2 | 24 | 2842 | ||
| GenBank mRNA | AK155448.1 | - | 24 | 2852 | ||
| AK169967.1 | - | 24 | 2870 | |||
| BC130260.1 | - | 6 | 816 | |||
| BC152337.1 | - | 6 | 816 | |||

Atp8b4
Genomic Map