siRNA Details
| Gene Symbol: Tgm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289747.1 | NP_001276676.1 | 11 | 1697 | ||
| NM_001289748.1 | NP_001276677.1 | 10 | 1534 | |||
| NM_001289749.1 | NP_001276678.1 | 8 | 1191 | |||
| NM_177726.4 | NP_808394.1 | 10 | 1553 | |||
| GenBank mRNA | AK037229.1 | - | 10 | 1543 | ||
| AY159126.1 | - | 10 | 1543 | |||
| AY177606.1 | - | 8 | 1181 | |||
| AY177607.1 | - | 10 | 1534 | |||
| BC112420.1 | - | 10 | 1530 | 40061385 | ||
| BC113796.1 | - | 11 | 1674 | 40061392 | ||

Tgm6
Genomic Map