siRNA Details
| Gene Symbol: Lzts3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291027.1 | NP_001277956.1 | 4 | 1150 | ||
| NM_001291028.1 | NP_001277957.1 | 4 | 808 | |||
| NM_197945.4 | NP_922936.3 | 4 | 1150 | |||
| GenBank mRNA | AK148251.1 | - | 4 | 1109 | ||
| AK161711.1 | - | 4 | 1104 | |||
| BC049892.1 | - | 2 | 711 | |||
| BC058280.1 | - | 3 | 732 | 6529102 | ||

Lzts3
Genomic Map