siRNA Details
| Gene Symbol: Fermt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198029.2 | NP_932146.2 | 15 | 2663 | ||
| GenBank mRNA | AF073489.1 | - | 12 | 1449 | ||
| AK030947.1 | - | 14 | 2853 | |||
| AK050804.1 | - | 14 | 1811 | |||
| BC029093.1 | - | 6 | 782 | |||

Fermt1
Genomic Map