siRNA Details
| Gene Symbol: L3mbtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081338.1 | NP_001074807.1 | 6, 7 | 874 | ||
| GenBank mRNA | AJ875399.1 | - | 5, 6 | 692 | ||
| AJ875400.1 | - | 4, 5 | 546 | |||
| AJ875401.1 | - | 5, 6 | 564 | |||
| AJ875402.1 | - | 2, 3 | 424 | |||
| AK049859.1 | - | 6, 7 | 868 | |||
| AK220350.1 | - | 4, 5 | 622 | |||
| BC116639.1 | - | 6, 7 | 938 | 40061282 | ||

L3mbtl1
Genomic Map