siRNA Details
| Gene Symbol: Slc7a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172861.3 | NP_766449.1 | 3 | 661 | ||
| GenBank mRNA | AK044448.1 | - | 3 | 654 | ||
| AK163358.1 | - | 3 | 705 | |||
| AK173217.1 | - | 3 | 637 | |||
| BC061928.1 | - | 3 | 637 | 5359765 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001134615.

Slc7a14
Genomic Map